Please login to your online account to display your discounted pricing

anti-&italic_begin;Clostridium difficile&italic_end; Toxin A, Clone: PCG4.1, Novus Biologicals™

$110.17 - $494.00

Specifications

Clone PCG4.1
Host Species Murine
Isotype κ
Immunogen Full length protein (&italic_begin;C. difficile&italic_end;).
Antigen Clostridium difficile Toxin A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001066SS

 
novus biologicals canada
NB6001066SS
Unlabeled 0.02mg Each for $110.17

NB6001066

 
novus biologicals canada
NB6001066
Unlabeled 1mg Each for $494.00
Description & Specifications

Specifications

Clone PCG4.1
Host Species Murine
Isotype κ
Immunogen Full length protein (&italic_begin;C. difficile&italic_end;).
Antigen Clostridium difficile Toxin A
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blot
Test Specificity C. difficile Toxin A. Does not cross react with C. difficile Toxin B. Cross-reacts with C. difficile. Does not cross react with C. difficile toxin B.
Formulation 10mM Sodium Phosphate, 7.4 containing 150mM Sodium Chloride
Concentration 1.16mg/mL