Please login to your online account to display your discounted pricing

anti-&italic_begin;Clostridium difficile&italic_end; Toxin A, Clone: PCG4.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001066SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001066SS

  • / Each
 

Description & Specifications

Specifications

Test Specificity C. difficile Toxin A. Does not cross react with C. difficile Toxin B. Cross-reacts with C. difficile. Does not cross react with C. difficile toxin B.
Quantity 0.02mg
Antigen Clostridium difficile Toxin A
Applications ELISA
Applications Western Blotting
Immunogen Full length protein (&italic_begin;C. difficile&italic_end;).
Isotype κ
Cross Reactivity Bacteria
Formulation 10mM Sodium Phosphate, 7.4 containing 150mM Sodium Chloride
Monoclonal or Polyclonal Monoclonal
Concentration 1.16mg/mL
Conjugate Unlabeled
Format Purified
Host Species Mouse
Regulatory Status RUO
Primary or Secondary Primary
Clone PCG4.1