Please login to your online account to display your discounted pricing

anti-CLEC9a, Clone: 14N8D7, Novus Biologicals™

$109.00 - $307.00

Specifications

Clone 14N8D7
Host Species Murine
Isotype κ
Immunogen A portion of amino acids 50-130 of human CLEC9A was used as the immunogen for the antibody. 60 74
Antigen CLEC9a
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227087

 
novus biologicals canada
NBP227087
Unlabeled 0.1mg Each for $307.00

NBP227087SS

 
novus biologicals canada
NBP227087SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 14N8D7
Host Species Murine
Isotype κ
Immunogen A portion of amino acids 50-130 of human CLEC9A was used as the immunogen for the antibody. 60 74
Antigen CLEC9a
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in Human, Orangutan, Chimp and Rhesus Monkey, 80% in Dog, pand and Horse, 73% in Bovine and 60% homologous in Mouse. CLEC9A is a glycoprotein visualized by western blot between ∽25-45 kD.
Gene Accession No. Q6UXN8
Gene ID (Entrez) 283420
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL