Please login to your online account to display your discounted pricing

anti-Cited-2, Clone: JA22, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone JA22
Host Species Murine
Isotype κ
Immunogen Human CITED 2 protein [UniProt No. Q99967]
Antigen Cited-2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100136

 
novus biologicals canada
NB100136
Unlabeled 0.1mL Each for $329.00

NB100136SS

 
novus biologicals canada
NB100136SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone JA22
Host Species Murine
Isotype κ
Immunogen Human CITED 2 protein [UniProt No. Q99967]
Antigen Cited-2
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q99967
Gene ID (Entrez) 10370