Please login to your online account to display your discounted pricing

anti-CIP2A, Clone: 2G10, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11059722

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11059722

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 57650
Test Specificity p90 autoantigen (2G10). Human, mouse, rat and primate. Other species have not been tested.
Formulation Tris-glycine, 150mM NaCl
Concentration 1.8mg/mL
Gene Accession No. Q8TCG1
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Format Purified
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Antigen CIP2A
Isotype κ
Quantity 0.1mL
Regulatory Status RUO
Host Species Mouse
Immunogen A synthetic peptide made to a C-terminal portion of the human CIP2A protein. [UniProt No. Q8TCG1]
Clone 2G10