Please login to your online account to display your discounted pricing

anti-Chromogranin A, Clone: PHE5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229427

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229427

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Regulatory Status RUO
Antigen Chromogranin A
Format Concentrated
Immunogen A recombinant protein fragment specific to Chromogranin A was used as the immunogen for the PHE5 antibody.
Isotype κ
Clone PHE5
Host Species Mouse
Formulation Culture Medium
Gene Accession No. P10645
Gene ID (Entrez) 1113
Primary or Secondary Primary
Conjugate Unlabeled
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Applications Immunohistochemistry (Paraffin)