Please login to your online account to display your discounted pricing

anti-Chromogranin A, Clone: PHE5, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone PHE5
Host Species Mouse
Isotype κ
Immunogen A recombinant protein fragment specific to Chromogranin A was used as the immunogen for the PHE5 antibody.
Antigen Chromogranin A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229427

 
novus biologicals canada
NBP229427
Unlabeled 0.5mL Each for $321.00

NBP2294271M

 
novus biologicals canada
NBP2294271ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone PHE5
Host Species Mouse
Isotype κ
Immunogen A recombinant protein fragment specific to Chromogranin A was used as the immunogen for the PHE5 antibody.
Antigen Chromogranin A
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Gene Accession No. P10645
Gene ID (Entrez) 1113
Formulation Culture Medium