Please login to your online account to display your discounted pricing

anti-Chromogranin A, Clone: PHE5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2294271ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2294271M

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Isotype κ
Quantity 1mL
Clone PHE5
Antigen Chromogranin A
Host Species Mouse
Immunogen A recombinant protein fragment specific to Chromogranin A was used as the immunogen for the PHE5 antibody.
Format Concentrated
Applications Immunohistochemistry (Paraffin)
Formulation Culture Medium
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene Accession No. P10645
Gene ID (Entrez) 1113
Regulatory Status RUO