Please login to your online account to display your discounted pricing

anti-Chromogranin A, Clone: LK2H10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232956

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232956

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Clone LK2H10
Formulation PBS with 0.05% BSA.
Gene Accession No. P10645
Gene ID (Entrez) 1113
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Immunogen Human pheochromocytoma
Test Specificity Guinea Pig (Negative), Rabbit (Negative), Sheep (Negative).
Antigen Chromogranin A