Please login to your online account to display your discounted pricing

anti-Chromogranin A, Clone: LK2H10, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone LK2H10
Host Species Mouse
Isotype κ
Immunogen Human pheochromocytoma
Antigen Chromogranin A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329561M

 
novus biologicals canada
NBP2329561ML
Unlabeled 1mL Each for $429.00

NBP232956

 
novus biologicals canada
NBP232956
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone LK2H10
Host Species Mouse
Isotype κ
Immunogen Human pheochromocytoma
Antigen Chromogranin A
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Guinea Pig (Negative), Rabbit (Negative), Sheep (Negative).
Gene Accession No. P10645
Gene ID (Entrez) 1113
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL