Please login to your online account to display your discounted pricing

anti-Chromogranin A, Clone: LK2H10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329561ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329561M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Clone LK2H10
Isotype κ
Quantity 1mL
Antigen Chromogranin A
Regulatory Status RUO
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 1113
Gene Accession No. P10645
Primary or Secondary Primary
Test Specificity Guinea Pig (Negative), Rabbit (Negative), Sheep (Negative).
Concentration 0.2mg/mL
Conjugate Unlabeled
Format Purified
Host Species Mouse
Immunogen Human pheochromocytoma
Monoclonal or Polyclonal Monoclonal