Please login to your online account to display your discounted pricing

anti-Chk1, Clone: 2G1D5, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 2G1D5
Host Species Mouse
Isotype IgG1
Immunogen Purified recombinant fragment of Chk1 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. O14757]
Antigen Chk1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001768

 
novus biologicals canada
NB1001768
Unlabeled 0.1mL Each for $309.05

NB1001768SS

 
novus biologicals canada
NB1001768SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 2G1D5
Host Species Mouse
Isotype IgG1
Immunogen Purified recombinant fragment of Chk1 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. O14757]
Antigen Chk1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. O14757
Gene ID (Entrez) 1111
Formulation PBS
Concentration 1.0mg/mL