Please login to your online account to display your discounted pricing

anti-CHAF1B, Clone: SS 53, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone SS 53
Host Species Murine
Isotype IgG1
Immunogen Recombinant human CHAF1B [UniProt No. Q13112]
Antigen CHAF1B
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500212

 
novus biologicals canada
NB500212
Unlabeled 0.1mL Each for $309.05

NB500212SS

 
novus biologicals canada
NB500212SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone SS 53
Host Species Murine
Isotype IgG1
Immunogen Recombinant human CHAF1B [UniProt No. Q13112]
Antigen CHAF1B
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q13112
Gene ID (Entrez) 8208