Please login to your online account to display your discounted pricing

anti-CHAF1A, Clone: SS 1 1-13, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone SS 1 1-13
Host Species Murine
Isotype IgG1
Immunogen Recombinant human CHAF1A [UniProt No. Q13111]
Antigen CHAF1A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500207

 
novus biologicals canada
NB500207
Unlabeled 0.2mL Each for $309.05

NB500207SS

 
novus biologicals canada
NB500207SS
Unlabeled 0.05mL Each for $109.00
Description & Specifications

Specifications

Clone SS 1 1-13
Host Species Murine
Isotype IgG1
Immunogen Recombinant human CHAF1A [UniProt No. Q13111]
Antigen CHAF1A
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q13111
Gene ID (Entrez) 10036