Please login to your online account to display your discounted pricing

anti-CFTR, Clone: CF3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300511SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300511SS

  • / Each
 

Description & Specifications

Specifications

Antigen CFTR
Applications Blocking
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blot
Clone CF3
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Ascites
Formulation Diluted ascites
Gene Accession No. P13569
Host Species Murine
Immunogen Synthetic peptide : G(103)RIIASYDPDNKEER(117). Immunizing peptide corresponds to amino acid residues 103-117 found in the first extracellular loop of human and rabbit CFTR. This sequence is highly conserved in mouse
Isotype IgM
Quantity 0.02mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1080
Test Specificity CFTR (CF3). Reported cross-reactivity in Rat (see product references). Not yet tested in other species.
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Rat