Please login to your online account to display your discounted pricing

anti-CEA, Clone: CEA31, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP233012

 View all options for this product


For Research Use Only

Catalog No. NBP233012


Description & Specifications


Antigen CEA
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone CEA31
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P06731
Host Species Mouse
Immunogen Human colon carcinoma extract
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal