Please login to your online account to display your discounted pricing

anti-CEA, Clone: CEA31, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone CEA31
Host Species Mouse
Isotype κ
Immunogen Human colon carcinoma extract
Antigen CEA
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233012

 
novus biologicals canada
NBP233012
Unlabeled 0.5mL Each for $321.00

NBP2330121M

 
novus biologicals canada
NBP2330121ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone CEA31
Host Species Mouse
Isotype κ
Immunogen Human colon carcinoma extract
Antigen CEA
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P06731
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL