Please login to your online account to display your discounted pricing

anti-CEA, Clone: CEA31, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2330121ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2330121M

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human colon carcinoma extract
Quantity 1mL
Host Species Mouse
Isotype κ
Clone CEA31
Cross Reactivity Human
Cross Reactivity Primate
Regulatory Status RUO
Primary or Secondary Primary
Conjugate Unlabeled
Concentration 0.2mg/mL
Gene Accession No. P06731
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Antigen CEA