Please login to your online account to display your discounted pricing

anti-CEA, Clone: C66/261, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2330101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2330101M

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Primate
Antigen CEA
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human recombinant CEA protein
Isotype κ
Quantity 1mL
Clone C66/261
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Gene Accession No. P06731
Primary or Secondary Primary
Regulatory Status RUO
Concentration 0.2mg/mL
Conjugate Unlabeled