Please login to your online account to display your discounted pricing

anti-CEA, Clone: C66/261, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233010

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233010

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Quantity 0.5mL
Antigen CEA
Host Species Mouse
Regulatory Status RUO
Format Purified
Clone C66/261
Cross Reactivity Human
Cross Reactivity Primate
Formulation PBS with 0.05% BSA.
Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. P06731
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Immunogen Human recombinant CEA protein