Please login to your online account to display your discounted pricing

anti-CEA, Clone: C66/261, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone C66/261
Host Species Mouse
Isotype κ
Immunogen Human recombinant CEA protein
Antigen CEA
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233010

 
novus biologicals canada
NBP233010
Unlabeled 0.5mL Each for $321.00

NBP2330101M

 
novus biologicals canada
NBP2330101ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone C66/261
Host Species Mouse
Isotype κ
Immunogen Human recombinant CEA protein
Antigen CEA
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P06731
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL