Please login to your online account to display your discounted pricing

anti-CDR2, Clone: 33, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP210509

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP210509

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-glycine, 150mM NaCl pH 7.5
Test Specificity Does not cross react with mouse Cdr2 and human Cdr3. Human. Does not react with Mouse.
Concentration 1.0mg/mL
Conjugate Unlabeled
Immunogen N-terminal MBP-tagged full length human CDR2 [Swiss-Prot No. Q01850]
Cross Reactivity Human
Isotype IgG1
Quantity 0.1mL
Regulatory Status RUO
Host Species Mouse
Format Purified
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CDR2
Monoclonal or Polyclonal Monoclonal
Clone 33
Primary or Secondary Primary
Gene Accession No. Q01850
Gene ID (Entrez) 1039