Please login to your online account to display your discounted pricing

anti-CDR2, Clone: 33, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 33
Host Species Murine
Isotype IgG1
Immunogen N-terminal MBP-tagged full length human CDR2 [Swiss-Prot No. Q01850]
Antigen CDR2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP210509

 
novus biologicals canada
NBP210509
Unlabeled 0.1mL Each for $309.05

NBP210509SS

 
novus biologicals canada
NBP210509SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 33
Host Species Murine
Isotype IgG1
Immunogen N-terminal MBP-tagged full length human CDR2 [Swiss-Prot No. Q01850]
Antigen CDR2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Does not cross react with mouse Cdr2 and human Cdr3. Human. Does not react with Mouse.
Gene Accession No. Q01850
Gene ID (Entrez) 1039
Formulation Tris-glycine, 150mM NaCl pH 7.5
Concentration 1.0mg/mL