Please login to your online account to display your discounted pricing

anti-CDR2, Clone: 33, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP210509SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP210509SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q01850
Quantity 0.025mL
Gene ID (Entrez) 1039
Immunogen N-terminal MBP-tagged full length human CDR2 [Swiss-Prot No. Q01850]
Regulatory Status RUO
Cross Reactivity Human
Isotype IgG1
Host Species Mouse
Format Purified
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CDR2
Formulation Tris-glycine, 150mM NaCl pH 7.5
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Does not cross react with mouse Cdr2 and human Cdr3. Human. Does not react with Mouse.
Clone 33
Concentration 1.0mg/mL
Conjugate Unlabeled