Please login to your online account to display your discounted pricing

anti-Cdc20, Clone: AR12, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232954

 View all options for this product


For Research Use Only

Catalog No. NBP232954


Description & Specifications


Antigen Cdc20
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone AR12
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q12834
Host Species Mouse
Immunogen Urea-denatured His6 Cdc20 human recombinant protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 991