Please login to your online account to display your discounted pricing

anti-Cdc20, Clone: AR12, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone AR12
Host Species Mouse
Isotype κ
Immunogen Urea-denatured His6 Cdc20 human recombinant protein
Antigen Cdc20
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232954

 
novus biologicals canada
NBP232954
Unlabeled 0.5mL Each for $321.00

NBP2329541M

 
novus biologicals canada
NBP2329541ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone AR12
Host Species Mouse
Isotype κ
Immunogen Urea-denatured His6 Cdc20 human recombinant protein
Antigen Cdc20
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. Q12834
Gene ID (Entrez) 991
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL