Please login to your online account to display your discounted pricing

anti-Cdc20, Clone: AR12, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329541ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329541M

  • / Each
 

Description & Specifications

Specifications

Antigen Cdc20
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone AR12
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q12834
Host Species Mouse
Immunogen Urea-denatured His6 Cdc20 human recombinant protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 991