Please login to your online account to display your discounted pricing

anti-CD90/Thy1, Clone: F15-42-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB60066302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB60066302

  • / Each
 

Description & Specifications

Specifications

Antigen CD90/Thy1
Isotype κ
Cross Reactivity Human
Cross Reactivity Primate
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen Purified human brain Thy1.
Format Purified
Clone F15-42-1
Gene Accession No. P04216
Primary or Secondary Primary
Test Specificity CD90 (F15-42-1). Cynomolgus Monkey.
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7070
Concentration 0.2mg/mL
Regulatory Status RUO
Quantity 0.2mg