Please login to your online account to display your discounted pricing

anti-CD90/Thy1, Clone: F15-42-1, Novus Biologicals™

$362.00 - $473.99

Specifications

Clone F15-42-1
Host Species Murine
Isotype κ
Immunogen Purified human brain Thy1.
Antigen CD90/Thy1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600663

 
novus biologicals canada
NB600663
Unlabeled 0.1mg Each for $362.00

NB60066302

 
novus biologicals canada
NB60066302MG
Unlabeled 0.2mg Each for $473.99
Description & Specifications

Specifications

Clone F15-42-1
Host Species Murine
Isotype κ
Immunogen Purified human brain Thy1.
Antigen CD90/Thy1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Test Specificity CD90 (F15-42-1). Cynomolgus Monkey.
Gene Accession No. P04216
Gene ID (Entrez) 7070
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL