Please login to your online account to display your discounted pricing

anti-CD9, Biotin, Clone: MEM-61, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081616

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081616

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Regulatory Status RUO
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Isotype IgG1
Clone MEM-61
Quantity 0.1mg
Primary or Secondary Primary
Formulation PBS [pH 7.4]
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 928
Test Specificity This recognizes an epitope on the second extracellular domain (EC2) of the human CD9 antigen.
Conjugate Biotin
Host Species Mouse
Concentration 1.0mg/mL
Antigen CD9
Immunogen Tissue/cell preparation (Human). (Pre-B line Nalm-6).