Please login to your online account to display your discounted pricing

anti-CD9, Biotin, Clone: MEM-61, Novus Biologicals™

$384.00 - $384.00

Specifications

Clone MEM-61
Host Species Murine
Isotype IgG1
Immunogen Tissue/cell preparation (Human). (Pre-B line Nalm-6).
Antigen CD9
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11081616

 
novus biologicals canada
NB11081616
Biotin 0.1mg Each for $384.00
Description & Specifications

Specifications

Clone MEM-61
Host Species Murine
Isotype IgG1
Immunogen Tissue/cell preparation (Human). (Pre-B line Nalm-6).
Antigen CD9
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This recognizes an epitope on the second extracellular domain (EC2) of the human CD9 antigen.
Gene ID (Entrez) 928
Formulation PBS [pH 7.4]
Concentration 1.0mg/mL