Please login to your online account to display your discounted pricing

anti-CD9, Biotin, Clone: MEM-61, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081616SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081616T

  • / Each
 

Description & Specifications

Specifications

Quantity 0.02mg
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1
Clone MEM-61
Regulatory Status RUO
Format Affinity Purified
Test Specificity This recognizes an epitope on the second extracellular domain (EC2) of the human CD9 antigen.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Conjugate Biotin
Gene ID (Entrez) 928
Formulation PBS [pH 7.4]
Host Species Mouse
Antigen CD9
Immunogen Tissue/cell preparation (Human). (Pre-B line Nalm-6).
Cross Reactivity Human