Please login to your online account to display your discounted pricing

anti-CD8 alpha, Clone: C8/468, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone C8/468
Host Species Mouse
Isotype IgG1
Immunogen Human CD8 recombinant protein
Antigen CD8 alpha
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329521M

 
novus biologicals canada
NBP2329521ML
Unlabeled 1mL Each for $429.00

NBP232952

 
novus biologicals canada
NBP232952
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone C8/468
Host Species Mouse
Isotype IgG1
Immunogen Human CD8 recombinant protein
Antigen CD8 alpha
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P01732
Gene ID (Entrez) 925
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL