Please login to your online account to display your discounted pricing

anti-CD8 alpha, Clone: C8/468, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329521ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329521M

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Isotype IgG1
Regulatory Status RUO
Quantity 1mL
Immunogen Human CD8 recombinant protein
Clone C8/468
Antigen CD8 alpha
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Gene Accession No. P01732
Primary or Secondary Primary
Gene ID (Entrez) 925
Conjugate Unlabeled
Host Species Mouse
Monoclonal or Polyclonal Monoclonal