Please login to your online account to display your discounted pricing

anti-CD8 alpha, Clone: C8/468, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232952

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232952

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Format Purified
Quantity 0.5mL
Host Species Mouse
Clone C8/468
Cross Reactivity Human
Isotype IgG1
Antigen CD8 alpha
Concentration 0.2mg/mL
Gene ID (Entrez) 925
Gene Accession No. P01732
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Conjugate Unlabeled
Primary or Secondary Primary
Immunogen Human CD8 recombinant protein