Please login to your online account to display your discounted pricing

anti-CD8 alpha, Clone: C8/144B, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328361ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328361M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Format Purified
Immunogen A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule.
Isotype IgG1
Quantity 1mL
Antigen CD8 alpha
Gene Accession No. P01732
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 925
Conjugate Unlabeled
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Host Species Mouse
Clone C8/144B
Cross Reactivity Human