Please login to your online account to display your discounted pricing

anti-CD8 alpha, Clone: C8/144B, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232836

 View all options for this product


For Research Use Only

Catalog No. NBP232836


Description & Specifications


Antigen CD8 alpha
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone C8/144B
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01732
Host Species Mouse
Immunogen A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule.
Isotype IgG1
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 925