Please login to your online account to display your discounted pricing

anti-CD8 alpha, Clone: C8/144B, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone C8/144B
Host Species Mouse
Isotype IgG1
Immunogen A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule.
Antigen CD8 alpha
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232836

 
novus biologicals canada
NBP232836
Unlabeled 0.5mL Each for $321.00

NBP2328361M

 
novus biologicals canada
NBP2328361ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone C8/144B
Host Species Mouse
Isotype IgG1
Immunogen A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule.
Antigen CD8 alpha
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P01732
Gene ID (Entrez) 925
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL