Please login to your online account to display your discounted pricing

anti-CD79A, Clone: JCB117, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229438

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229438

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone JCB117
Quantity 0.5mL
Host Species Mouse
Immunogen 1994)].
Cross Reactivity Human
Regulatory Status RUO
Formulation Culture Medium
Conjugate Unlabeled
Gene Accession No. P11912
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 973
Test Specificity A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells. This complex of mb-1 and B29 polypeptides and immunoglobulin constitute the B cell Ag receptor. CD79a first appears at pre B cell stage, early in maturation, and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Anti-CD79a is generally used to complement anti-CD20 especially for mature B-cell lymphomas after treatment with Rituximab (anti-CD20). This antibody will stain many of the same lymphomas as anti-CD20, but also is more likely to stain B-lymphoblastic lymphoma/leukemia than is anti-CD20. Anti-CD79a also stains more cases of plasma cell myeloma and occasionally some types of endothelial cells as well.
Format Concentrated
Antigen CD79A