Please login to your online account to display your discounted pricing

anti-CD79A, Clone: JCB117, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone JCB117
Host Species Mouse
Isotype κ
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Antigen CD79A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329491M

 
novus biologicals canada
NBP2329491ML
Unlabeled 1mL Each for $429.00

NBP232949

 
novus biologicals canada
NBP232949
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone JCB117
Host Species Mouse
Isotype κ
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Antigen CD79A
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P11912
Gene ID (Entrez) 973
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL