Please login to your online account to display your discounted pricing

anti-CD79A, Clone: JCB117, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone JCB117
Host Species Mouse
Isotype κ
Immunogen 1994)].
Antigen CD79A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2294381M

 
novus biologicals canada
NBP2294381ML
Unlabeled 1mL Each for $429.00

NBP229438

 
novus biologicals canada
NBP229438
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone JCB117
Host Species Mouse
Isotype κ
Immunogen 1994)].
Antigen CD79A
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells. This complex of mb-1 and B29 polypeptides and immunoglobulin constitute the B cell Ag receptor. CD79a first appears at pre B cell stage, early in maturation, and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Anti-CD79a is generally used to complement anti-CD20 especially for mature B-cell lymphomas after treatment with Rituximab (anti-CD20). This antibody will stain many of the same lymphomas as anti-CD20, but also is more likely to stain B-lymphoblastic lymphoma/leukemia than is anti-CD20. Anti-CD79a also stains more cases of plasma cell myeloma and occasionally some types of endothelial cells as well.
Gene Accession No. P11912
Gene ID (Entrez) 973
Formulation Culture Medium