Please login to your online account to display your discounted pricing

anti-CD79A, Clone: JCB117, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329491ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329491M

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Regulatory Status RUO
Clone JCB117
Host Species Mouse
Isotype κ
Formulation PBS with 0.05% BSA.
Quantity 1mL
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Gene Accession No. P11912
Primary or Secondary Primary
Gene ID (Entrez) 973
Conjugate Unlabeled
Antigen CD79A
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.