Please login to your online account to display your discounted pricing

anti-CD79A, Clone: JCB117, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232949

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232949

  • / Each
 

Description & Specifications

Specifications

Antigen CD79A
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone JCB117
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P11912
Host Species Mouse
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 973