Please login to your online account to display your discounted pricing

anti-CD68/SR-D1, Clone: SPM281, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP1-74570SS

 View all options for this product


For Research Use Only

Catalog No. NBP174570SS


Description & Specifications


Antigen CD68/SR-D1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM281
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation This affinity purified antibody is supplied in sterile Phosphate buffered saline (pH 7.2) containing antibody stabilizer.
Gene Accession No. P34810
Host Species Mouse
Immunogen Subcellular fraction of human alveolar macrophages
Isotype κ
Quantity 0.02mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 968
Test Specificity This antibody recognizes ∽110 kDa of human CD68 protein. It cross reacts with human, mouse, rat and monkey. The other species are not tested.