Please login to your online account to display your discounted pricing

anti-CD68/SR-D1, Clone: SPM281, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP174570

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP174570

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Quantity 0.1mg
Regulatory Status RUO
Antigen CD68/SR-D1
Clone SPM281
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 968
Test Specificity This antibody recognizes ∽110 kDa of human CD68 protein. It cross reacts with human, mouse, rat and monkey. The other species are not tested.
Conjugate Unlabeled
Formulation This affinity purified antibody is supplied in sterile Phosphate buffered saline (pH 7.2) containing antibody stabilizer.
Gene Accession No. P34810
Concentration 0.5mg/mL
Host Species Mouse
Immunogen Subcellular fraction of human alveolar macrophages