Please login to your online account to display your discounted pricing

anti-CD68/SR-D1, Clone: SPM281, Novus Biologicals™

$109.00 - $375.00

Specifications

Clone SPM281
Host Species Mouse
Isotype κ
Immunogen Subcellular fraction of human alveolar macrophages
Antigen CD68/SR-D1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP174570

 
novus biologicals canada
NBP174570
Unlabeled 0.1mg Each for $375.00

NBP174570SS

 
novus biologicals canada
NBP174570SS
Unlabeled 0.02mg Each for $109.00
Description & Specifications

Specifications

Clone SPM281
Host Species Mouse
Isotype κ
Immunogen Subcellular fraction of human alveolar macrophages
Antigen CD68/SR-D1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This antibody recognizes ∽110 kDa of human CD68 protein. It cross reacts with human, mouse, rat and monkey. The other species are not tested.
Gene Accession No. P34810
Gene ID (Entrez) 968
Formulation This affinity purified antibody is supplied in sterile Phosphate buffered saline (pH 7.2) containing antibody stabilizer.
Concentration 0.5mg/mL