Please login to your online account to display your discounted pricing

anti-CD68/SR-D1, Clone: KP1, Novus Biologicals™

$164.00 - $384.00

Specifications

Clone KP1
Host Species Murine
Isotype κ
Immunogen Subcellular fraction of human alveolar macrophages.
Antigen CD68/SR-D1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100683

 
novus biologicals canada
NB100683
Unlabeled 0.5mL Each for $384.00

NB100683SS

 
novus biologicals canada
NB100683SS
Unlabeled 0.125mL Each for $164.00
Description & Specifications

Specifications

Clone KP1
Host Species Murine
Isotype κ
Immunogen Subcellular fraction of human alveolar macrophages.
Antigen CD68/SR-D1
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This is specific to macrophages in a wide variety of human tissues. It reacts with myeloid precursors and peripheral blood granulocytes. It also stains a cell population known as Plasmacytoid T cells. Human and Mouse. Rat reactivity reported in scientific literature (PMID: 25058444).
Gene Accession No. P34810
Gene ID (Entrez) 968