Please login to your online account to display your discounted pricing

anti-CD68/SR-D1, Clone: KP1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100683SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100683SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P34810
Test Specificity This is specific to macrophages in a wide variety of human tissues. It reacts with myeloid precursors and peripheral blood granulocytes. It also stains a cell population known as Plasmacytoid T cells. Human and Mouse. Rat reactivity reported in scientific literature (PMID: 25058444).
Gene ID (Entrez) 968
Isotype κ
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.125mL
Antigen CD68/SR-D1
Clone KP1
Immunogen Subcellular fraction of human alveolar macrophages.
Format Supernatant
Host Species Mouse