Please login to your online account to display your discounted pricing

anti-CD68/SR-D1, Clone: KP1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100683

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100683

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 968
Gene Accession No. P34810
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Test Specificity This is specific to macrophages in a wide variety of human tissues. It reacts with myeloid precursors and peripheral blood granulocytes. It also stains a cell population known as Plasmacytoid T cells. Human and Mouse. Rat reactivity reported in scientific literature (PMID: 25058444).
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype κ
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Subcellular fraction of human alveolar macrophages.
Format Supernatant
Regulatory Status RUO
Quantity 0.5mL
Clone KP1
Antigen CD68/SR-D1
Host Species Mouse