Please login to your online account to display your discounted pricing

anti-CD68/SR-D1, Clone: C68/684, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone C68/684
Host Species Mouse
Isotype κ
Immunogen Subcellular fraction of human alveolar macrophages
Antigen CD68/SR-D1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328321M

 
novus biologicals canada
NBP2328321ML
Unlabeled 1mL Each for $429.00

NBP232832

 
novus biologicals canada
NBP232832
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone C68/684
Host Species Mouse
Isotype κ
Immunogen Subcellular fraction of human alveolar macrophages
Antigen CD68/SR-D1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P34810
Gene ID (Entrez) 968
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL